Learn More
C&G Containers 500 mL Natural HDPE Oblong Bottle, High Impact Resistance, Space-Saving Design
SDP
Catalog No. NC3662443
Encompass_Preferred
500 mL Natural HDPE Oblong bottle is designed for laboratory applications, providing a robust moisture barrier and high impact resistance. Its space-saving shape optimizes benchtop organization and transport, making it ideal for storing and mixing chemical solutions. Engineered for chemical compatibility, this BPA-free bottle is leakproof and certified, ensuring safe handling during various scientific processes.
Catalog No. NC3662443 Supplier C&G Containers Supplier No. LPV016220602
500 mL Natural HDPE Oblong bottle is designed for laboratory applications, providing a robust moisture barrier and high impact resistance. Its space-saving shape optimizes benchtop organization and transport, making it ideal for storing and mixing chemical solutions. Engineered for chemical compatibility, this BPA-free bottle is leakproof and certified, ensuring safe handling during various scientific processes.