Learn More
CELLTREAT Scientific Products 500mL Bottle Top Filter, 0.45m Nylon Filter, 75mm Diameter, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229733
Encompass_Preferred
CELLTREAT Scientific Products 500mL Nylong Bottle Top Filter. Easy-to-read molded graduations marks on bottle top filter. Bold easy-to-read printed membrane and pore size for easy identification. Curved bottle design for easy handling. Printed lot number and expiration date on every bottle top. Improved filter support provides faster flow rates. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping. 0.45m PES membrane ideal for solution clarification. Nylon (PA) membrane has excellent chemical compatibility and does not require pre-wetting. Bottle top filters fit on CELLTREAT solution bottles or other standard 45mm threaded solution storage bottles. Bottle top filters do not include solution bottles. Large diameter filter increases flow and prevents clogging. Membrane diameter 75mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 24/case.
Catalog No. 50-114-9105