Learn More
Boston Bioproducts Inc Paraformaldehyde (4%, PBS) - 500ml
Supplier:  Boston Bioproducts Inc BM155500ML
Encompass_Preferred
Paraformaldehyde solution is a neutral fixative solution used to fix tissues and cells in immunohistochemistry and immunocytochemistry protocols. Stabilized in a phosphate buffer for delivery, this reagent actively cross-links proteins and DNA, immobilizing cellular components and maintaining the integrity of cell morphology. This reagent supports the accurate visualization of cells and tissues under a microscope, ensuring that researchers can observe fine details without degradation.
Catalog No. NC9099744