Learn More
Chemglass Life Sciences 500mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 32 Thread
SDP
Catalog No. CG186115
Encompass_Preferred
500mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186115 Supplier Chemglass Life Sciences Supplier No. CG186115
500mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D. Vessels are available in sizes other than those listed. Please call for details.