Learn More
CELLTREAT Scientific Products 500mL Erlenmeyer Flask, Vent Cap, Plain Bottom, PETG, Sterile
SDP
Catalog No. 501656699
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Available with either vent or solid cap to prevent common mistakes made with combination caps such as sealing the cap completely and preventing gas exchange. Vent caps with over-sized vent area feature 0.22¿m hydrophobic membrane which allows for improved gas exchange without contamination. Avoid contamination with single-use, pre-sterilized flasks. Manufactured from medical grade virgin PETG which reduces gas permeability. Lightweight material is easier to handle than glass. Bottles include molded graduation lines. Individually bagged for easy storage and handling. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 12/case.
Catalog No. 50-165-6699 Supplier CELLTREAT Scientific Products Supplier No. 229809
Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Available with either vent or solid cap to prevent common mistakes made with combination caps such as sealing the cap completely and preventing gas exchange. Vent caps with over-sized vent area feature 0.22¿m hydrophobic membrane which allows for improved gas exchange without contamination. Avoid contamination with single-use, pre-sterilized flasks. Manufactured from medical grade virgin PETG which reduces gas permeability. Lightweight material is easier to handle than glass. Bottles include molded graduation lines. Individually bagged for easy storage and handling. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 12/case.