Learn More
Triforest Enterprises 500ML sterile PETG square media bottle with standard cap and heat-shrink band around closure and neck for tamper-evident seal
Catalog No. 50607667
Encompass_Preferred
TriForest Square PETG media bottles are ideal for storage of sterile reagents. Bottles are pre-sterilized, SAL 10-6, and include a standard cap with heat-shrink band around closure and neck for tamper-evident seal. Each tray is packaged for sterile production with multi-barrier packaging.
Catalog No. 50-607-667 Supplier Triforest Enterprises Supplier No. BGC0500SB
TriForest Square PETG media bottles are ideal for storage of sterile reagents. Bottles are pre-sterilized, SAL 10-6, and include a standard cap with heat-shrink band around closure and neck for tamper-evident seal. Each tray is packaged for sterile production with multi-barrier packaging.