Learn More
Ace Glass, Inc. 500ML PLASTIC COATING 500ML PLASTIC COATING
SDP
Supplier:  Ace Glass, Inc. 1310010
Encompass_Preferred
500mL jar of plastic coating. This easy-to-apply plastic coating forms a protective film around the outside of a glass component, and will normally contain both the glass particles and the materials inside the vessel in case of breakage during an experiment. This coating is a hard, adherent film, and is resistant to leaching and bleed-out in a bath. The coating will withstand -20 to 121 degrees C for extended periods of time without degradation. Do not use in direct flame. Direction for applying and curing supplied.
Catalog No. 50-983-478