Learn More
Ace Glass, Inc. 500ML POR A BOTTLE 500ML POR A BOTTLE
SDP
Catalog No. 50974141
Encompass_Preferred
500mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-141 Supplier Ace Glass, Inc. Supplier No. 716670
500mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.