Learn More
Ace Glass, Inc. 500mL porosity A bottle complete
SDP
Catalog No. 50979118
Encompass_Preferred
500mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D. Includes stopper.
Catalog No. 50-979-118 Supplier Ace Glass, Inc. Supplier No. 716622
500mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D. Includes stopper.