Learn More
Ace Glass, Inc. 500mL gas wash stopper only, porosity A
SDP
Catalog No. 50974107
Encompass_Preferred
500mL gas wash stopper with porosity A (145-174micron) fritted disc.
Catalog No. 50-974-107 Supplier Ace Glass, Inc. Supplier No. 716280
500mL gas wash stopper with porosity A (145-174micron) fritted disc.