Learn More
Ace Glass, Inc. 500ML POR B BOTTLE 500ML POR B BOTTLE
SDP
Catalog No. 50979120
Encompass_Preferred
500mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-979-120 Supplier Ace Glass, Inc. Supplier No. 716672
500mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.