Learn More
Ace Glass, Inc. 500mL complete gas washing bottle, porosity B
SDP
Supplier:  Ace Glass, Inc. 716224
Encompass_Preferred
500mL complete gas washing bottle with a 30mm fritted porosity B (70-100micron) disc.
Catalog No. 50-979-106