Learn More
Ace Glass, Inc. 500mL porosity B bottle complete
SDP
Catalog No. 50974136
Encompass_Preferred
500mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D. Includes stopper.
Catalog No. 50-974-136 Supplier Ace Glass, Inc. Supplier No. 716624
500mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D. Includes stopper.