Learn More
Ace Glass, Inc. 500mL gas wash stopper only, porosity B
SDP
Catalog No. 50974108
Encompass_Preferred
500mL gas wash stopper with porosity B (70-100micron) fritted disc.
Catalog No. 50-974-108 Supplier Ace Glass, Inc. Supplier No. 716282
500mL gas wash stopper with porosity B (70-100micron) fritted disc.