Learn More
Ace Glass, Inc. 500ML POR C BOTTLE 500ML POR C BOTTLE
SDP
Catalog No. 50974142
Encompass_Preferred
500mL gas washing bottle with porosity C (25-50 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-142 Supplier Ace Glass, Inc. Supplier No. 716674
500mL gas washing bottle with porosity C (25-50 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.