Learn More
Ace Glass, Inc. 500mL complete gas washing bottle, porosity C
SDP
Supplier:  Ace Glass, Inc. 716226
Encompass_Preferred
500mL complete gas washing bottle with a 30mm fritted porosity C (25-50micron) disc.
Catalog No. 50-974-100