Learn More
Ace Glass, Inc. 500mL gas wash stopper only, porosity C
SDP
Catalog No. 50974120
Encompass_Preferred
500mL gas washing stopper with porosity C (25-50 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-120 Supplier Ace Glass, Inc. Supplier No. 716384
500mL gas washing stopper with porosity C (25-50 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.