Learn More
Ace Glass, Inc. 500mL gas wash bottle "C"
SDP
Catalog No. 50974137
Encompass_Preferred
500mL tall form gas washing bottle with "C" porosity fritted disc and 29/42 stopper.
Catalog No. 50-974-137 Supplier Ace Glass, Inc. Supplier No. 716626
500mL tall form gas washing bottle with "C" porosity fritted disc and 29/42 stopper.