Learn More
Foxx Life Sciences 500mL Narrow Mouth PP Reagent Bottles, 48CS
GREENER_CHOICE
Catalog No. 5000846669
Encompass_Preferred
EZLabpure™ 500mL PP Reagent Bottles Autoclavable, Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth PP Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. PP Chemical Reagent Bottles are rigid and fully autoclavable without deforming making them durable and reliable for storing aqueous laboratory solutions.
Features
Autoclavable up to 121°C
PP Storage Reagent Bottles are rigid and translucent for easy content visibility.
PP Storage Bottles offer excellent resistance to aqueous solutions and non-corrosive chemicals.
Leak-proof Linerless screw caps for secure sealing.
PP Bottles with Low Liquid Retention
Catalog No. 50-008-46669 Supplier Foxx Life Sciences Supplier No. 7651G21FLS
EZLabpure™ 500mL PP Reagent Bottles Autoclavable, Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these lightweight, shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth PP Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. PP Chemical Reagent Bottles are rigid and fully autoclavable without deforming making them durable and reliable for storing aqueous laboratory solutions.
Features
Autoclavable up to 121°C
PP Storage Reagent Bottles are rigid and translucent for easy content visibility.
PP Storage Bottles offer excellent resistance to aqueous solutions and non-corrosive chemicals.
Leak-proof Linerless screw caps for secure sealing.
PP Bottles with Low Liquid Retention