Learn More
Chemglass Life Sciences Flask, Round Bottom, 500mL, Heavy Wall, 3-Neck, 29/42 CN - 29/42 SN's Angled 20°
SDP
Catalog No. CG152441
Encompass_Preferred
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 29/42
Catalog No. CG152441 Supplier Chemglass Life Sciences Supplier No. CG152441
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 29/42