Learn More
Chemglass Life Sciences 500mL 2-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 1-SN 24/40 Outer
SDP
Catalog No. CG151806
Encompass_Preferred
500mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.
Catalog No. CG151806 Supplier Chemglass Life Sciences Supplier No. CG151806
500mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.