Learn More
Chemglass Life Sciences Flask, Round Bottom, 500mL, Heavy Wall, 3-Neck, 29/42 CN - 24/40 SNs, Vertical
SDP
Catalog No. CG152502
Encompass_Preferred
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Vertical. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and side necks. Indentations increase agitation of the contents of the flask during stirring.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. CG152502 Supplier Chemglass Life Sciences Supplier No. CG152502
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Vertical. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and side necks. Indentations increase agitation of the contents of the flask during stirring.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum