Learn More
Chemglass Life Sciences 500ml. Reaction Vessel, Cylindrical, Morton, 100mm Schott O-Ring Flange, 105mm O.D. x 135mm Height
SDP
Catalog No. CG192201
Encompass_Preferred
500ml. Reaction Vessel, Cylindrical, Morton, 100mm Schott O-Ring Flange, 105mm O.D. x 135mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Indentations increase agitation of the contents of the vessel during stirring. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. CG192201 Supplier Chemglass Life Sciences Supplier No. CG192201
500ml. Reaction Vessel, Cylindrical, Morton, 100mm Schott O-Ring Flange, 105mm O.D. x 135mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Indentations increase agitation of the contents of the vessel during stirring. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum