Learn More
Chemglass Life Sciences 500mL Single Neck Evaporating Flask, 24/25 Outer Joint, Recovery Flask
SDP
Catalog No. CG151225
Encompass_Preferred
500mL Single Neck Evaporating Flask, 24/25 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. CG151225 Supplier Chemglass Life Sciences Supplier No. CG151225
500mL Single Neck Evaporating Flask, 24/25 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.