Learn More
Chemglass Life Sciences Chemglass OptiTherm 500 mL Recovery Flask Insert (for OPT1000 block)
SDP
Catalog No. 502733538
Encompass_Preferred
The Chemglass OptiTherm 500 mL recovery flask insert is an anodized berry aluminum accessory engineered to support 500 mL round-bottom recovery flasks within compatible reaction blocks. It improves thermal contact between the flask and the block for efficient heat transfer during distillation, reflux, and solvent recovery applications, and provides a secure, low-profile fit to minimize flask movement.
  • Anodized aluminum construction for improved heat transfer and corrosion resistance.
  • Designed to support 500 mL round-bottom recovery flasks.
  • Low-profile design maximizes contact with the heating surface.
  • Color-coded anodized finish for quick identification.
  • Compatible with large-format reaction and heating blocks used for recovery and distillation.
Catalog No. 50-273-3538 Supplier Chemglass Life Sciences Supplier No. CG1992R500
The Chemglass OptiTherm 500 mL recovery flask insert is an anodized berry aluminum accessory engineered to support 500 mL round-bottom recovery flasks within compatible reaction blocks. It improves thermal contact between the flask and the block for efficient heat transfer during distillation, reflux, and solvent recovery applications, and provides a secure, low-profile fit to minimize flask movement.
  • Anodized aluminum construction for improved heat transfer and corrosion resistance.
  • Designed to support 500 mL round-bottom recovery flasks.
  • Low-profile design maximizes contact with the heating surface.
  • Color-coded anodized finish for quick identification.
  • Compatible with large-format reaction and heating blocks used for recovery and distillation.