Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 500mL round bottom flasks, max diameter 3.98 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately
SDP
Catalog No. 50977361
Encompass
Combo mantle, 3 position, low temperature, fits 500mL round bottom flasks, max diameter 3.98 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately
Catalog No. 50-977-361 Supplier Ace Glass, Inc. Supplier No. 12061108
Combo mantle, 3 position, low temperature, fits 500mL round bottom flasks, max diameter 3.98 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately