Learn More
Chemglass Life Sciences Separatory Funnel, Squibb Pear-Shaped, 500 mL, PTFE Stopcock
SDP
Catalog No. NC2099117
Encompass_Preferred
500 mL Squibb (pear-shaped) separatory funnel for laboratory liquid-liquid extraction and phase separation. The funnel features a chemically resistant PTFE stopcock and is supplied with a #27 flask-length stopper. The PTFE plug in the stopcock barrel helps reduce freezing and grease contamination.
  • 500 mL capacity suitable for lab-scale liquid-liquid extractions.
  • Pear-shaped (Squibb) body for clear phase separation.
  • PTFE stopcock for chemical resistance and low contamination.
  • Supplied with a #27 flask-length stopper.
  • Borosilicate glass construction for chemical and thermal resistance.
This item is not returnable. View return policy
Catalog No. NC2099117 Supplier Chemglass Life Sciences Supplier No. CG8622500
500 mL Squibb (pear-shaped) separatory funnel for laboratory liquid-liquid extraction and phase separation. The funnel features a chemically resistant PTFE stopcock and is supplied with a #27 flask-length stopper. The PTFE plug in the stopcock barrel helps reduce freezing and grease contamination.
  • 500 mL capacity suitable for lab-scale liquid-liquid extractions.
  • Pear-shaped (Squibb) body for clear phase separation.
  • PTFE stopcock for chemical resistance and low contamination.
  • Supplied with a #27 flask-length stopper.
  • Borosilicate glass construction for chemical and thermal resistance.