Learn More
Ace Glass, Inc. Complete assembly, #25 Ace-Thred top, includes 500mL flask, #25 well, septum adapter, #7 & #25 Ace-Thred nylon bushings, 8mm septa for side port, 24/40 septa for top, FETFE O-rings
SDP
Catalog No. 50978154
Encompass_Preferred
Catalog No. 50-978-154 Supplier Ace Glass, Inc. Supplier No. 1420540