Learn More
CELLTREAT Scientific Products 500mL Media Bottle, Square, PETG, Sterile
SDP
Catalog No. NC2119220
Encompass_Preferred
Ideal for media prep and storage of laboratory reagents and biological buffers. Square design saves valuable shelf space. High clarity for easy viewing of contents. Plastic bottles are a shatterproof alternative to glass media bottlesPETG material reduces gas permeability. Molded graduations on two sides. Flat surface polyethylene (HDPE) caps are ideal for marking.
This item is not returnable. View return policy
Catalog No. NC2119220 Supplier CELLTREAT Scientific Products Supplier No. 229788
Ideal for media prep and storage of laboratory reagents and biological buffers. Square design saves valuable shelf space. High clarity for easy viewing of contents. Plastic bottles are a shatterproof alternative to glass media bottlesPETG material reduces gas permeability. Molded graduations on two sides. Flat surface polyethylene (HDPE) caps are ideal for marking.