Learn More
Eagle Stainless Container Eagle Stainless, Biological Transfer Bottle, 0.5 Liter, 316L Stainless Steel. (BTB Series)
SDP
Catalog No. NC0479771
Encompass_Preferred
0.5 Liter Biological Transfer Bottle with 2" tri-clamp opening, type 316L stainless steel. Supplied with 2" stainless steel endcap, 2" platinum cured silicone gasket and 2" heavy duty clamp. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC0479771 Supplier Eagle Stainless Container Supplier No. BTB7316LA
0.5 Liter Biological Transfer Bottle with 2" tri-clamp opening, type 316L stainless steel. Supplied with 2" stainless steel endcap, 2" platinum cured silicone gasket and 2" heavy duty clamp. Pharmaceutical grade finish of 10 Ra or better.