Learn More
Chemglass Life Sciences Stopper Only, 500mL Gas Washing Bottle
SDP
Catalog No. 501531010
Encompass_Preferred
Stopper Only, 500mL Gas Washing Bottle. Stopper is held in place with two CG-110-02, 18mm Stainless Steel springs. Springs are supplied when bottle is ordered complete. Image shown is the complete bottle.
Catalog No. 50-153-1010 Supplier Chemglass Life Sciences Supplier No. CG111208
Stopper Only, 500mL Gas Washing Bottle. Stopper is held in place with two CG-110-02, 18mm Stainless Steel springs. Springs are supplied when bottle is ordered complete. Image shown is the complete bottle.