Learn More
Chemglass Life Sciences 500mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 190mm OAH
SDP
Catalog No. AF009404
Encompass_Preferred
500mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 190mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. AF009404 Supplier Chemglass Life Sciences Supplier No. AF009404
500mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 190mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.