Learn More
CELLTREAT Scientific Products 500mL Suspension Culture Flask - Vent Cap, Sterile
SDP
Catalog No. NC3796182
Encompass_Preferred
Stable base enables filling with flask standing uprightConvenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membraneLarge opening, canted neck design and moderate angles allow easy access to entire flask surface Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks onlyFlask is printed with lot number and expiration date for easy identification and tracking150cm2 Flask features U-shaped design for more efficient cell harvesting"
Catalog No. NC3796182 Supplier CELLTREAT Scientific Products Supplier No. 229550
Stable base enables filling with flask standing uprightConvenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membraneLarge opening, canted neck design and moderate angles allow easy access to entire flask surface Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks onlyFlask is printed with lot number and expiration date for easy identification and tracking150cm2 Flask features U-shaped design for more efficient cell harvesting"