Learn More
S2 Media 500mL Tryptic Soy Broth with 1% Polysorbate 80 (TSB w/T), 500mL PC Media Bottle and Septum Cap, 1/each
Catalog No. NC3900355
Encompass_Preferred
Tryptic Soy Broth with 1% Polysorbate 80 (TSB w/T) with a septum cap is a non-selective and non-differential general growth medium used in for the detection and recovery of microorganisms, particularly in the presence of antimicrobial agents. The addition of Polysorbate 80 (Tween 80) serves as a neutralizer, inactivating residual disinfectants or preservatives that could otherwise inhibit microbial growth. This formulation is frequently employed in sterility testing, environmental monitoring, disinfectant efficacy studies, and microbial limits testing of non-sterile products to ensure accurate detection of contaminants.• Product base meets AOAC, BAM, EP, ISO, SM-Dairy, SMWW, USDA, and USP-NF reference methods and performance criteria.• TSB should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Septum Cap
Catalog No. NC3900355 Supplier S2 Media Supplier No. 5527
Tryptic Soy Broth with 1% Polysorbate 80 (TSB w/T) with a septum cap is a non-selective and non-differential general growth medium used in for the detection and recovery of microorganisms, particularly in the presence of antimicrobial agents. The addition of Polysorbate 80 (Tween 80) serves as a neutralizer, inactivating residual disinfectants or preservatives that could otherwise inhibit microbial growth. This formulation is frequently employed in sterility testing, environmental monitoring, disinfectant efficacy studies, and microbial limits testing of non-sterile products to ensure accurate detection of contaminants.• Product base meets AOAC, BAM, EP, ISO, SM-Dairy, SMWW, USDA, and USP-NF reference methods and performance criteria.• TSB should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Septum Cap