Learn More
CELLTREAT Scientific Products 500mL Tube Cell Strainer, 100μm, Yellow, Individually Wrapped, Sterile, Case Quantity 50
SDP
Catalog No. 5000670034
Encompass_Preferred
Large diameter 500mL Cell Strainers are designed for use with 500mL Centrifuge Tubes (229463)
Accommodates tubes with inner neck diameters between 35 and 40 mm
Can be used for many applications, including isolating cells and removing clumps or debris.
Hydrophilic treatment increases flow and reduces the effects of surface tension
Polypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materials
Extended lip on housing enables aseptic handling with forcep.
Elongated gripping tab on housing allows for easy handling
Exclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers
Catalog No. 50-006-70034 Supplier CELLTREAT Scientific Products Supplier No. 230495
Large diameter 500mL Cell Strainers are designed for use with 500mL Centrifuge Tubes (229463)
Accommodates tubes with inner neck diameters between 35 and 40 mm
Can be used for many applications, including isolating cells and removing clumps or debris.
Hydrophilic treatment increases flow and reduces the effects of surface tension
Polypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materials
Extended lip on housing enables aseptic handling with forcep.
Elongated gripping tab on housing allows for easy handling
Exclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers