Learn More
CELLTREAT Scientific Products 500mL Tube Cell Strainer, 70μm, White, Individually Wrapped, Sterile
SDP
Catalog No. 5000144346
Encompass_Preferred
Large diameter 500mL Cell Strainers are designed for use with 500mL Centrifuge Tubes (229463).Accommodates tubes with inner neck diameters between 35 and 40mmCan be used for many applications including isolating cells and removing clumps or debrisHydrophilic treatment increases flow and reduces the effects of surface tensionPolypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingExclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers
Catalog No. 50-001-44346 Supplier CELLTREAT Scientific Products Supplier No. 230493
Large diameter 500mL Cell Strainers are designed for use with 500mL Centrifuge Tubes (229463).Accommodates tubes with inner neck diameters between 35 and 40mmCan be used for many applications including isolating cells and removing clumps or debrisHydrophilic treatment increases flow and reduces the effects of surface tensionPolypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingExclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers