Learn More
Chemglass Life Sciences 500mL 3-Neck Round Bottom Flask (RBF) , Half Jacket, 1-CN 29/42 Outer, 2-SN 24/40 Outer, 6mm Drain Valve
SDP
Catalog No. CG153902
Encompass_Preferred
500mL 3-Neck RBF, Half Jacket, 1-CN 29/42 Outer, 2-SN 24/40 Outer, 6mm Drain Valve. Lower half of flask has a jacket permitting heating or cooling of contents with fluid from a constant temperature bath. Design of the Chem-Drain valve incorporates a PTFE plunger that allows contents of flask to drain directly through the bore of the plunger. When closed, the tip of the PTFE plug seals into the lower radius of the flask. This permits total mixing & drainage of the flask's contents. The Chem-Drain is 6mm. This dimension refers to the straight section of the plunger. When open, the contents of the flask drain through three openings into the plunger's main bore. Lower portion of the plunger has a 1/2in section for attachment of flexible tubing. The 6mm size allows drainage of solids to 1.5mm. Supplied standard in a three-neck configuration. Valve size is as listed but flasks with capacity of 2 Liters or more can be fitted with the 12mm valve. See CG-510 for replacement Chem-drain plugs.
Catalog No. CG153902 Supplier Chemglass Life Sciences Supplier No. CG153902
500mL 3-Neck RBF, Half Jacket, 1-CN 29/42 Outer, 2-SN 24/40 Outer, 6mm Drain Valve. Lower half of flask has a jacket permitting heating or cooling of contents with fluid from a constant temperature bath. Design of the Chem-Drain valve incorporates a PTFE plunger that allows contents of flask to drain directly through the bore of the plunger. When closed, the tip of the PTFE plug seals into the lower radius of the flask. This permits total mixing & drainage of the flask's contents. The Chem-Drain is 6mm. This dimension refers to the straight section of the plunger. When open, the contents of the flask drain through three openings into the plunger's main bore. Lower portion of the plunger has a 1/2in section for attachment of flexible tubing. The 6mm size allows drainage of solids to 1.5mm. Supplied standard in a three-neck configuration. Valve size is as listed but flasks with capacity of 2 Liters or more can be fitted with the 12mm valve. See CG-510 for replacement Chem-drain plugs.