Learn More
Chemglass Life Sciences Flask, Round Bottom, 500mL, Heavy Wall, 3-Neck, 29/42 CN - 24/40 SNs, Vertical, Jacketed
SDP
Catalog No. CG153802
Encompass_Preferred
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Vertical, Jacketed. Round bottom, HEAVY WALL, three-neck flask with standard taper outer joints on the center and side necks. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid.
Catalog No. CG153802 Supplier Chemglass Life Sciences Supplier No. CG153802
Flask, Round Bottom, 500mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Vertical, Jacketed. Round bottom, HEAVY WALL, three-neck flask with standard taper outer joints on the center and side necks. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid.