Learn More
Teknova 500mM EDTA, pH 8.0. 1000mL, Endotoxin Tested, Sterile. 8 pack.
SDP
Catalog No. 502387569
Encompass
EDTA (ethylenediaminetetraacetic acid) is a chelating agent used to remove metal ions from solutions and inhibit the growth of microorganisms. EDTA Solution is an essential component of many laboratory procedures, and it can be used in a variety of settings, including research laboratories, hospitals, and clinics.
Catalog No. 50-238-7569 Supplier Teknova Supplier No. E03118PK
EDTA (ethylenediaminetetraacetic acid) is a chelating agent used to remove metal ions from solutions and inhibit the growth of microorganisms. EDTA Solution is an essential component of many laboratory procedures, and it can be used in a variety of settings, including research laboratories, hospitals, and clinics.