Learn More
Teknova 500mM EDTA, pH 8.0. 1000mL, Sterile.
SDP
Catalog No. 50841660
Encompass
EDTA (ethylenediaminetetraacetic acid) is a chelating agent used to remove metal ions from solutions and inhibit the growth of microorganisms. EDTA Solution is an essential component of many laboratory procedures, and it can be used in a variety of settings, including research laboratories, hospitals, and clinics.
This item is not returnable. View return policy
Catalog No. 50-841-660 Supplier Teknova Supplier No. E0308
EDTA (ethylenediaminetetraacetic acid) is a chelating agent used to remove metal ions from solutions and inhibit the growth of microorganisms. EDTA Solution is an essential component of many laboratory procedures, and it can be used in a variety of settings, including research laboratories, hospitals, and clinics.