Learn More
Boston Bioproducts Inc 50 mM Sodium Phosphate, 2 mM EDTA (pH 7.0) - 2x1L
Catalog No. 5000330295
Encompass_Preferred
Bioprocess Grade - 50 mM Sodium Phosphate 2 mM EDTA (pH 7 0) - 2x1L (For Research Use and Further Manufacturing Only)
Catalog No. 50-003-30295 Supplier Boston Bioproducts Inc Supplier No. BPS3312X1L
Bioprocess Grade - 50 mM Sodium Phosphate 2 mM EDTA (pH 7 0) - 2x1L (For Research Use and Further Manufacturing Only)