Learn More
Biolegend Anti-CD171 (L1CAM)
Supplier:  Biolegend 838101
Encompass_Preferred
Anti-CD171 (L1CAM) [74-5H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC, IP, IHC-P; Size: 500 uL
Catalog No. NC1782644