Learn More
Perkin Elmer US LLC Type C0.5 MEINHARD Glass Nebulizer for NexION 300/350
Catalog No. NC0349517
Encompass_Preferred
MEINHARD™ Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.
Catalog No. NC0349517 Supplier Perkin Elmer US LLC Supplier No. N8145012
MEINHARD™ Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.