Learn More
SAI Infusion Technologies 500uL Lithium Heparin Blood Tube
SDP
Catalog No. NC0891576
Encompass
500uL Lithium Heparin Blood Tube for collection of blood via drip or transfer. Featuring a slim Easy Cap which is removed with a small twist and replaced a small push. Skirted tube has a false bottom for easy pipetting. Optimized tube geometry ensures thorough sample mixing. 100/box. Product is for laboratory animal research only.
Catalog No. NC0891576 Supplier SAI Infusion Technologies Supplier No. MVH500
500uL Lithium Heparin Blood Tube for collection of blood via drip or transfer. Featuring a slim Easy Cap which is removed with a small twist and replaced a small push. Skirted tube has a false bottom for easy pipetting. Optimized tube geometry ensures thorough sample mixing. 100/box. Product is for laboratory animal research only.