Learn More
Ace Glass, Inc. 500 microliter gas-tight, removable needle syringe
SDP
Catalog No. 50972634
Encompass
500 microliter gas-tight syringe with a removable needle (RN), and a 3/4 length, bevel point style #2 needle.
Catalog No. 50-972-634 Supplier Ace Glass, Inc. Supplier No. 593313
500 microliter gas-tight syringe with a removable needle (RN), and a 3/4 length, bevel point style #2 needle.