Learn More
Bellco Glass, Inc. 500µL Pipette, MLA, Precision, Yellow
SDP
Catalog No. 502330464
Encompass_Preferred
500µL Pipette, MLA, Precision, YellowTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorYellowPrimary MaterialAluminum Alloy & Stainless SteelVolume500 μLVistaLab Tip Fit SizeLarge 1250μLUniversal FitNoAccuracy±1.00%Precision0.40%
Catalog No. 50-233-0464 Supplier Bellco Glass, Inc. Supplier No. 451900009
500µL Pipette, MLA, Precision, YellowTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorYellowPrimary MaterialAluminum Alloy & Stainless SteelVolume500 μLVistaLab Tip Fit SizeLarge 1250μLUniversal FitNoAccuracy±1.00%Precision0.40%