Learn More
SAI Infusion Technologies 500uL Serum Separator Gel Blood Tube
SDP
Catalog No. NC0831084
Encompass
500uL Serum Gel Blood Tube with Clotting Activator and serum separator gel. Featuring a slim Easy Cap which is removed with a small twist and replaced a small push. Skirted tube has a false bottom for easy pipetting. Optimized tube geometry ensures thorough sample mixing with Clotting Activator. 100/box. Product is for laboratory animal research only.
Catalog No. NC0831084 Supplier SAI Infusion Technologies Supplier No. MVSG500
500uL Serum Gel Blood Tube with Clotting Activator and serum separator gel. Featuring a slim Easy Cap which is removed with a small twist and replaced a small push. Skirted tube has a false bottom for easy pipetting. Optimized tube geometry ensures thorough sample mixing with Clotting Activator. 100/box. Product is for laboratory animal research only.