Learn More
Trajan Scientific VICI Valco Sample Loop, 500l, Cheminert, Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and Ferrules (VAL.CSL500)
Catalog No. 50635011
Encompass
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL500 (NEW LPP PN: VAL.CSL500). Thermo Part Number: CH-952936. Made in the USA.
Catalog No. 50-635-011 Supplier Trajan Scientific Supplier No. PAL.CSL500
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL500 (NEW LPP PN: VAL.CSL500). Thermo Part Number: CH-952936. Made in the USA.