Learn More
Trajan Scientific Hamilton 500L Syringe, FN, 22, PTFE, Pt. 2, 51mm
Catalog No. 50711785
Encompass_Preferred
Hamilton Syringe, 500μL, Hamilton Syringe, Fixed Needle, Gas Tight, 22 Gauge, NDL Length 51mm, Pt. 3, Plunger Tip PTFE, Single
Catalog No. 50-711-785 Supplier Trajan Scientific Supplier No. HAM.81216
Hamilton Syringe, 500μL, Hamilton Syringe, Fixed Needle, Gas Tight, 22 Gauge, NDL Length 51mm, Pt. 3, Plunger Tip PTFE, Single