Learn More
Zymo Research Corporation 500x Concentrated Arabinose Inducer (1 ml)
SDP
Catalog No. 501957631
Encompass
A 1 ml vial is sufficient for 500 ml of XJ Autolysis E. coli culture.
Catalog No. 50-195-7631 Supplier Zymo Research Corporation Supplier No. A20011
A 1 ml vial is sufficient for 500 ml of XJ Autolysis E. coli culture.